Tubes without Cap, Round Base, PS
With green label/print, 5 ml, 12 mm, 75 mm, Non graduated, BAGA
Catalog # 77587-880
Supplier: SARSTEDT INC
: 62.476.326
Jump to:
Product Details & Documents
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, this range offers the right product for every laboratory and analytical requirement.
- Material: PS
- Base shape: Round base
- Amount of packaging reduced by 40% (if StackPack option is used)
- Polystyrene (PS) tubes for maximum transparency
- Different push caps for a variety of applications
- Volumes from 3.5 to 26 ml
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.
Specifications
- Ø:12 mm
- Volume:5 ml
- Description:With green label/print
- Length:75 mm
- Packaging:BAGA
- Graduations:Non graduated



