Orders LinkContactUsLinkComponent
Colour-Coded Inserts
Colour-Coded Inserts
Black
Catalog # 77588-168
Supplier:   SARSTEDT INC
Supplier #:  65.713.010
Colour-Coded Inserts
Catalog # 77588-168
Supplier:  SARSTEDT INC
Colour-Coded Inserts
Colour-Coded Inserts
Black
Price
$17.19
Log in for availability
Pack of 500 Case of 5000
Product Details & Documents
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Polypropylene material
  • Different colors for better coding
  • Easy to insert
  • Suitable for screw caps 65.712
This variety of color-coded inserts allows for quick and flexible coding and identification.
Specifications
  • Color:
    Black