HYDRO SM QAS STD 10X0.4G VIALS
MSPP-CRM3150
Frequently Bought Together
Immobiline™ DryStrip Gels, Cytiva
: Cytiva
Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
: G-Biosciences
ELISA KIT ARG8 VASOPRESSINS
: ARBOR ASSAYS MS
RBC Lysis Buffer, Zymo Research
: Zymo Research
Rat Fibronectin ELISA Kit, Rockland Immunochemicals
: Rockland Immunochemical
FGF-BASIC (154AA) HUMAN >95% 1MG
: Genscript
DNA Elution Buffer, Zymo Research
: Zymo Research
MARKER MAURICE CIEF PI-8.40 LYOPH 210UL
: PROTEIN SIMPLE MS
Western Blot Boxes
: Alkali Scientific
Buffer P1 (Red), Zymo Research
: Zymo Research
Y-Lysis Buffer, Zymo Research
: Zymo Research
Zyppy Elution Buffer, Zymo Research
: Zymo Research
INHIBITOR YEAST 100X PROTEASEARREST 5ML
: G-Biosciences
PI MARKER ICE280/ICE3 6.61 200UL
: PROTEIN SIMPLE MS
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
: New England Biolabs (NEB)
CASTING TRAY FOR ELECTROPHORESIS SYSTEM
: G-Biosciences
PINPOINT BINDING BUFFER 6 ML F/ISOLATION
: Zymo Research
AGAROSE ALFA SELECTOR ST 2ML
: NANOTAG BIOTECHNOLOGIES GMBH MS
ScanLater™ Western Blot Assay Kit, Molecular Devices
: Molecular Devices
TrueBlot® Protein A Magnetic Beads, Rockland Immunochemicals
: Rockland Immunochemical