Orders LinkContactUsLinkComponent
Mini Drums
Mini Drums
6 L, Stainless steel 304, 175 mm, 280 mm
Catalog # 77577-974
Supplier #:  A434-6
Mini Drums
Catalog # 77577-974
Mini Drums
Mini Drums
6 L, Stainless steel 304, 175 mm, 280 mm
Price
$505.00
Log in for availability
Each
Product Details & Documents
Mini drums make for ideal storage and production containers.
  • Made from high-quality 304 stainless steel
  • Ideal for both powders and liquids
  • Supplied with a drop-on lid
  • Toggle clamps and O-Ring seals are optional extras
  • Available from stock
  • Fast delivery
This range of mini drums are ideal for the storage of both powders and liquids. They come complete with a drop-on lid. Optional extras include toggle clamps and O-Ring seals, enabling the lid to be firmly secured.
Specifications
  • Ø:
    175 mm
  • Capacity:
    6 L
  • Height:
    280 mm
  • Material:
    Stainless steel 304
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 10 L
    Silicone seal for mini-drum, 10 L
    77578-226
    Silicone
    Log in for availability
    Each - $48.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 3 L
    Silicone seal for mini-drum, 3 L
    77578-240
    Silicone
    Log in for availability
    Each - $38.10
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 15 L
    Silicone seal for mini-drum, 15 L
    77578-230
    Silicone
    Log in for availability
    Each - $63.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for storage container, 20 L
    Silicone seal for storage container, 20 L
    77578-234
    Silicone
    Log in for availability
    Each - $63.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 1 L
    Silicone seal for mini-drum, 1 L
    77578-232
    Silicone
    Log in for availability
    Each - $21.70
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 12 L
    Silicone seal for mini-drum, 12 L
    77578-228
    Silicone
    Log in for availability
    Each - $53.60
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 8 L
    Silicone seal for mini-drum, 8 L
    77578-248
    Silicone
    Log in for availability
    Each - $40.80
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for storage container, 50 L
    Silicone seal for storage container, 50 L
    77578-242
    Silicone
    Log in for availability
    Each - $62.80
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 2 L
    Silicone seal for mini-drum, 2 L
    77578-238
    Silicone
    Log in for availability
    Each - $33.60
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for storage container, 25 L
    Silicone seal for storage container, 25 L
    77578-236
    Silicone
    Log in for availability
    Each - $63.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 5 L
    Silicone seal for mini-drum, 5 L
    77578-244
    Silicone
    Log in for availability
    Each - $39.10
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 6 L
    Silicone seal for mini-drum, 6 L
    77578-246
    Silicone
    Log in for availability
    Each - $38.40