Pointed Scissors
310 mm
Catalog # 77577-894
Supplier: SAMPLING SYSTEMS LTD
Supplier #: A364-300
Jump to:
Product Details & Documents
Surgical grade scissors for cutting.
- Made from surgical grade 420 stainless steel
- Great for cutting and manipulating samples
These high quality pointed scissors are made from surgical grade 420 stainless steel and great for cutting and manipulating.
Specifications
- Length:310 mm