Order Entry
United States
Orders LinkContactUsLinkComponent
SteriWare® Tablet Counting Triangles

SteriWare® Tablet Counting Triangles

Sterile, 230×230×13 mm, White
Catalog # 77576-970
Supplier #:  8066A-1S
SteriWare® Tablet Counting Triangles
Catalog # 77576-970
Supplier:  
SteriWare® Tablet Counting Triangles
SteriWare® Tablet Counting Triangles

Sterile, 230×230×13 mm, White

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
TABLET COUNTING TRIANGLE - PS. STERILE Catalog #78117-123

Product 78117-123 is a replacement for 77576-970 but varies by Brand, Packaging or Quantity.

Product Details & Documents

The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.

  • Made from FDA conforming PS
  • BSE/TSE free
  • Suitable for left handed and right handed operators
  • With an embossed scale to aid operation

Certifications: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.

Packaging: Individually bagged in a heat-sealed laminate bag.

Specifications
  • Color:
    White
  • Description:
    Sterile
  • W×D×H:
    230×230×13 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.