Order Entry
United States
Orders LinkContactUsLinkComponent
Measuring Scoops

Measuring Scoops

25 ml, 199 mm, 35 mm
Catalog # 77576-336
Supplier #:  1900A-25
Measuring Scoops
Catalog # 77576-336
Measuring Scoops
Measuring Scoops

25 ml, 199 mm, 35 mm

Price
$141.00
Log in for availability
Each
Product Details & Documents

These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.

  • Crevice-free
  • Polished
Specifications
  • Ø:
    35 mm
  • Capacity:
    25 ml
  • Length:
    199 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.