Order Entry
United States
Orders LinkContactUsLinkComponent
Measuring Scoops

Measuring Scoops

10 ml, 193 mm, 22 mm
Catalog # 77576-334
Supplier #:  1900A-10
Measuring Scoops
Catalog # 77576-334
Measuring Scoops
Measuring Scoops

10 ml, 193 mm, 22 mm

Price
$131.00
Log in for availability
Each
Product Details & Documents

These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.

  • Crevice-free
  • Polished
Specifications
  • Ø:
    22 mm
  • Capacity:
    10 ml
  • Length:
    193 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.