Order Entry
United States
Orders LinkContactUsLinkComponent
Sub Sampler Pro Bomb Sampler

Sub Sampler Pro Bomb Sampler

Bomb samplers, Nickel plated brass, 500 ml
Catalog # 77576-532
Supplier #:  5072A-500
Sub Sampler Pro Bomb Sampler
Catalog # 77576-532
Sub Sampler Pro Bomb Sampler
Sub Sampler Pro Bomb Sampler

Bomb samplers, Nickel plated brass, 500 ml

Price
$1,230.00
Log in for availability
Each
Product Details & Documents

Essential for flammable liquids as a heavy-duty sampler.

  • Metallic parts manufactured from stainless steel
  • Unique double action to sample from a set distance
  • Can be plunged until the assembly touches the bottom of the container

Specially designed for the environment throughout the world for both chemical and environmental sampling.

Certifications: Conforms to FDA CFR 177.2600.

Specifications
  • Capacity:
    500 ml
  • Description:
    Bomb samplers
  • Material:
    Nickel plated brass