Order Entry
United States
Orders LinkContactUsLinkComponent
 

TeleScoop Sampler for Liquids

Telescopic rod with PTFE clutch, Stainless steel 316L, 150 - 300 cm
Catalog # 77576-526
 :  5065S-3000
TeleScoop Sampler for Liquids
Catalog # 77576-526
TeleScoop Sampler for Liquids
TeleScoop Sampler for Liquids

Telescopic rod with PTFE clutch, Stainless steel 316L, 150 - 300 cm

Price
$921.00
Log in for availability
Each
Product Details & Documents

These telescopic handles can be used when sampling from overhead gantries or difficult to reach places.

  • Strong telescopic handle
  • Range of volumes and styles of collecting vessels

Suitable for taking samples from liquids and suspensions.

Specifications
  • Description:
    Telescopic rod with PTFE clutch
  • Length:
    150 - 300 cm
  • Material:
    Stainless steel 316L