Order Entry
United States
Orders LinkContactUsLinkComponent
 

Bottle Liquid Sampler

Bottle liquid sampler with 500 ml glass bottle, 500 ml, 1000 mm
  77576-438
 :  5031S-1000
Bottle Liquid Sampler
  77576-438
Bottle Liquid Sampler

Bottle liquid sampler with 500 ml glass bottle, 500 ml, 1000 mm

 
$2,100.00
   
Each
 

Incorporates a spring loaded mechanism, which means that liquid only flows in when the handle is squeezed allowing samples to be taken from set locations. Sampling Systems™ bottle liquid sampler takes samples straight into the bottle.

  • 316L Stainless Steel sampler body
  • Glass bottle
  • Single point
  • Sampling for low to medium viscosity liquids
  • Ideal for low volume (unit dose) sampling

The sampler is available with a cover that stops the bottle getting wet.

 
  • Capacity:
    500 ml
  • Description:
    Bottle liquid sampler with 500 ml glass bottle
  • Length:
    1000 mm