Order Entry
United States
Orders LinkContactUsLinkComponent
 

Dipping Bottles

Brass, 1000 ml, 300 mm, 75 mm
  77576-560
 :  5332B-1000
Dipping Bottles
  77576-560
Dipping Bottles
Dipping Bottles

Brass, 1000 ml, 300 mm, 75 mm

 
$318.00
   
Each
 

Dipping bottles are excellent for sampling in open water, as well as in tanks and drums.

  • Wide range available
  • Stainless steel 304 or brass construction
  • Customisable

Sampling liquids at depth is easy with these dipping bottles. Available in either 304 stainless steel or brass construction, they can take liquid samples from a variety of locations including open water kegs, drums, and wells.

 
  • Ø:
    75 mm
  • Capacity:
    1000 ml
  • Length:
    300 mm
  • Material:
    Brass
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.