Order Entry
United States
Orders LinkContactUsLinkComponent
 

Sampling Tips for UD Sleeve Sampler

Sampling tip for UD sleeve sampler, Stainless steel 316L, 1.4 ml
  77576-288
 :  1500W-14
Sampling Tips for UD Sleeve Sampler
  77576-288
Sampling Tips for UD Sleeve Sampler
Sampling Tips for UD Sleeve Sampler

Sampling tip for UD sleeve sampler, Stainless steel 316L, 1.4 ml

 
$145.00
   
Each
 

The UD Sleeve sampler is perfect for small volume powder sampling.

  • Extensively used in the pharmaceutical industry
  • Accurate sampling of free-flowing powders
  • Extension rods are avialble to increase the length of the sampler

The UD Sleeve Sampler is ideal for accurate sampling of small volumes of free-flowing powders. It is made from 316L stainless steel and available with a range of tips, enabling samples sizes between 0,1 and 2,5 ml. Various accessories are available, such as extension rods which anable the operator to extend the length of the sampler.

 
  • Volume:
    1.4 ml
  • Description:
    Sampling tip for UD sleeve sampler
  • Material:
    Stainless steel 316L