Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions

pH Buffer Solutions

pH buffer standard solution with certificate of analysis, 125 ml, 4.01
Catalog # 104088-604
Marketsource
Supplier:   Hach
Supplier #:  LZW9460.99
pH Buffer Solutions
Catalog # 104088-604
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions
pH Buffer Solutions

pH buffer standard solution with certificate of analysis, 125 ml, 4.01

CAS Number:  
Price
$28.37
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    pH buffer standard solution with certificate of analysis, 125 ml
  • pH:
    4.01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.