Order Entry
United States
Orders LinkContactUsLinkComponent
Buffer Solutions for Hardness Testing

Buffer Solutions for Hardness Testing

Catalog # 104088-328
Marketsource
Supplier:   Hach
Supplier #:  42449
Buffer Solutions for Hardness Testing
Catalog # 104088-328
Supplier:  Hach
Marketsource
CAS Number:  
Buffer Solutions for Hardness Testing
Buffer Solutions for Hardness Testing
CAS Number:  
Price
$59.74
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

This buffer is used for hardness and chelant determination by titration.

Documents

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.