Orders LinkContactUsLinkComponent
TNTplus™ Test Kits, COD
250 - 15000 mg/L
Catalog # 104088-660
Marketsource
Supplier:   Hach
 :  TNT82306
TNTplus™ Test Kits, COD
Catalog # 104088-660
Supplier:  Hach
Marketsource
CAS Number:  
TNTplus™ Test Kits, COD
TNTplus™ Test Kits, COD
250 - 15000 mg/L
CAS Number:  
Price
$702.00
Log in for availability
Each
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
Specifications
Specifications
  • Range:
    250 - 15000 mg/L