Order Entry
United States
Orders LinkContactUsLinkComponent
PH DO METER HQ4100 CDC

PH DO METER HQ4100 CDC

Catalog # 104088-532
Marketsource
Supplier:   Hach
Supplier #:  LEV015.53.41004
PH DO METER HQ4100 CDC
Catalog # 104088-532
Supplier:  Hach
Marketsource
CAS Number:  
PH DO METER HQ4100 CDC
CAS Number:  
Price
$3,857.54
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.