Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions

pH Buffer Solutions

pH buffer standard solution with certificate of analysis, 125 ml, 7
Catalog # 104088-606
Marketsource
Supplier:   Hach
Supplier #:  LZW9461.97
pH Buffer Solutions
Catalog # 104088-606
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions
pH Buffer Solutions

pH buffer standard solution with certificate of analysis, 125 ml, 7

CAS Number:  
Price
$28.37
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    pH buffer standard solution with certificate of analysis, 125 ml
  • pH:
    7

Specifications