Sample Cuvettes
1" / 25 mm, Glass
104088-228
Marketsource
: Hach
: 2495408
Sample cuvettes for photometric measurements.
- Available with or without lid
- Available reusable (made from glass) or for single use (made from plastics)
- Window material:Glass
- Pathlength:1" / 25 mm