TPH Reagent Sets
Chlorine (free), DPD, 0.02 - 2 mg/L, -, 10 ml, PP
Supplier: Hach
Supplier #: 2105569
Jump to:
Product Details & Documents
For the determination of total petroleum hydrocarbons (TPH) in soil and water.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Documents
Certificates
Search by Lot Numberghs_2774300
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Chemical method:DPD
- Description:Chlorine (free)
- Range:0.02 - 2 mg/L
- Type:PP
- Sample Size:10 ml
- No. of tests:-
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



