Orders LinkContactUsLinkComponent
TNTplus™ Test Kits, Phosphorus
TNTplus™ Test Kits, Phosphorus
Phosphorus TNT plus kit, reactive and total, UHR, Ascorbic acid, 2 - 20 mg/L
Catalog # 104088-286
Marketsource
Supplier:   Hach
Supplier #:  2742545
TNTplus™ Test Kits, Phosphorus
Catalog # 104088-286
Supplier:  Hach
Marketsource
CAS Number:  
TNTplus™ Test Kits, Phosphorus
TNTplus™ Test Kits, Phosphorus
Phosphorus TNT plus kit, reactive and total, UHR, Ascorbic acid, 2 - 20 mg/L
CAS Number:  
Price
$122.88
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
TNT Plus kit is used for the efficient and accurate determination of reactive (ortho) and total phosphorus (phosphate) using TNTplus® vials.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
Documents
Specifications
Specifications
  • Chemical method:
    Ascorbic acid
  • Description:
    Phosphorus TNT plus kit, reactive and total, UHR
  • Range:
    2 - 20 mg/L