Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffer Powder Pillows

±0.02, 25 °C, Pack of 20, 4.01 and 7.00, Red/yellow
Catalog # 104088-198
Marketsource
Supplier:   Hach
 :  2299264
Buffer Powder Pillows
Catalog # 104088-198
Supplier:  Hach
Marketsource
CAS Number:  
Buffer Powder Pillows
Buffer Powder Pillows

±0.02, 25 °C, Pack of 20, 4.01 and 7.00, Red/yellow

CAS Number:  
Price
$32.05
Log in for availability
Pack of 20
 
Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

 : The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Specifications

Specifications

  • Color:
    Red/yellow
  • Packed:
    Pack of 20
  • Reference temperature:
    25 °C
  • pH:
    4.01 and 7.00
  • pH accuracy:
    ±0.02

Specifications