Orders LinkContactUsLinkComponent
TPH Reagent Sets
TPH Reagent Sets
Chromium total set, Hypobromite oxidation, 0.01 - 0.70 mg/L, 100, -, PP
Catalog # 104088-186
Marketsource
Supplier:   Hach
Supplier #:  2242500
TPH Reagent Sets
Catalog # 104088-186
Supplier:  Hach
Marketsource
CAS Number:  
TPH Reagent Sets
TPH Reagent Sets
Chromium total set, Hypobromite oxidation, 0.01 - 0.70 mg/L, 100, -, PP
CAS Number:  
Price
$368.31
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
For the determination of total petroleum hydrocarbons (TPH) in soil and water.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Documents
Specifications
Specifications
  • Chemical method:
    Hypobromite oxidation
  • Description:
    Chromium total set
  • Range:
    0.01 - 0.70 mg/L
  • Type:
    PP
  • Sample Size:
  • No. of tests:
    100