Selenium 99+%, powder
Product Details & Documents
- CAS Number:7782-49-2
- Formula:Se
- MDL Number:MFCD00134090
- Molecular Weight:78.96 g/mol
- Boiling Pt:∼685 °C (1013 hPa)
- Melting Pt:217 °C
- Density:4.81 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-957-4
- UN:3283
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:Dark grey to black Powder
- Assay:≥99.0 %
- Residue after ignition:≤0.5 %
- Copper (Cu):≤100 ppm
- Iron (Fe):≤500 ppm
- Lead (Pb):≤500 ppm
- Nickel (Ni):≤100 ppm
- Sulfur (S):≤500 ppm
Specifications
- Pack type:Glass bottle
- Size:500 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



