Order Entry
United States
Orders LinkContactUsLinkComponent
Selenium 99+%, powder

Selenium 99+%, powder

Glass bottle, 100 g
Catalog # TS419271000
Toxic
Health
Supplier #:  419271000
Selenium 99+%, powder
Catalog # TS419271000
CAS Number:  7782-49-2
Selenium 99+%, powder

Glass bottle, 100 g

CAS Number:  7782-49-2
Price
$113.00
Log in for availability
Each(100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.81 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • UN:3283
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    Dark grey to black Powder
  • Assay:
    ≥99.0 %
  • Residue after ignition:
    ≤0.5 %
  • Copper (Cu):
    ≤100 ppm
  • Iron (Fe):
    ≤500 ppm
  • Lead (Pb):
    ≤500 ppm
  • Nickel (Ni):
    ≤100 ppm
  • Sulfur (S):
    ≤500 ppm

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.