Order Entry
United States
Orders LinkContactUsLinkComponent
 

Gram's decolourizing solution

Plastic bottle, 250 ml
Catalog # 77540-230
Supplier:   G-Biosciences
 :  786-2086
Gram's decolourizing solution
Catalog # 77540-230
Supplier:  G-Biosciences
CAS Number:  
Gram's decolourizing solution

Plastic bottle, 250 ml

CAS Number:  
Price
$26.15
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Gram Decolorizer is a mixture of 25% Acetone and 75% Isopropanol solution.

  • An aqueous solution of Acetone and Isopropanol

It is used for rapid decolorization before counterstaining

Synonyms:  Gram's Decolorizer Solution, Gram's Decoloriser Solution
  • MDL Number:MFCD00240361
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.