Dissolution Sampling Cannulas
Glass syringe, 20 cc
Catalog # AG12-3230
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
: 12-3230
Jump to:
Product Details & Documents
Available in two different lengths based on the volume of media; best used with full flow filters.
- For manual sampling, the Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters
- The sampling cannulas are available two in different lengths based on the volume of media: 500 or 900 ml
- An adjustable gauge can be included with this assembly to ensure repeatable positioning for each sample
The Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters. The sampling cannulas are available in two different lengths based on the volume of media being sampled from.
Specifications
- Description:Glass syringe, 20 cc