Order Entry
United States
Orders LinkContactUsLinkComponent
solution 100 mg/mL

solution 100 mg/mL

1 ml
Catalog # 77528-960
Supplier:   Teknova
Supplier #:  C8001
solution 100 mg/mL
Catalog # 77528-960
Supplier:  Teknova
CAS Number:  4697-36-3
solution 100 mg/mL

1 ml

CAS Number:  4697-36-3
Price
$429.76
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Carbenicillin is a semi-synthetic ampicillin analog, and like ampicillin is a beta-lactam antibiotic. Carbenicillin is a member of the carboxypenicillin subgroup of the penicillins.

It is effective against Gram-negative organisms including H. influenzae and E. coli. Carbenicillin is also used in transformed Agrobacter species. Carbenicillin was the first penicillin with significant activity against Pseudomonas species.

  • CAS Number:4697-36-3
  • Formula:C₁₇H₁₈N₂O₆S

Documents

Specifications

Specifications

  • Size:
    1 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.