BROTH 2XYT WITH 2% GLUCOSE STERILE 500ML
Catalog # 77510-100
Supplier: Teknova
BROTH 2XYT WITH 2% GLUCOSE STERILE 500ML
Catalog # 77510-100
Supplier: Teknova
Supplier Number: Y0175
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Gorilla Glue® Super Glue
Supplier: Janitorial Supplies
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
Supplier: Hardy Diagnostics
ONETAQ® 2X Master Mix with Standard Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
2XYT BROTH WITH 20% GLUCOSE 1000MLST
Supplier: Teknova