Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER AAV AEX AAV2 EQUILIBRATION 20630

BUFFER AAV AEX AAV2 EQUILIBRATION 20630

Catalog # 77509-416
Supplier:   Teknova
Supplier #:  20630
BUFFER AAV AEX AAV2 EQUILIBRATION 20630
Catalog # 77509-416
Supplier:  Teknova
CAS Number:  
BUFFER AAV AEX AAV2 EQUILIBRATION 20630
CAS Number:  
Price
$180.71
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.