Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER AAV AEX AAV2 EQUILIBRATION 20618

BUFFER AAV AEX AAV2 EQUILIBRATION 20618

Catalog # 77509-412
Supplier:   Teknova
Supplier #:  20618
BUFFER AAV AEX AAV2 EQUILIBRATION 20618
Catalog # 77509-412
Supplier:  Teknova
CAS Number:  
BUFFER AAV AEX AAV2 EQUILIBRATION 20618
CAS Number:  
Price
$144.57
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents