Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER SM STERILE 1000ML

BUFFER SM STERILE 1000ML

Catalog # 77509-998
Supplier:   Teknova
Supplier #:  S2210
BUFFER SM STERILE 1000ML
Catalog # 77509-998
Supplier:  Teknova
CAS Number:  
BUFFER SM STERILE 1000ML
CAS Number:  
Price
$182.20
Log in for availability
Each
Minimum order quantity is 2
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.