MEDIUM RPMI 1640 W/O-L-GLUTAMINE 500ML
Catalog # MSPP-9160500ML
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
MEDIUM RPMI 1640 W/O-L-GLUTAMINE 500ML
Catalog # MSPP-9160500ML
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Supplier Number: 9160-500ML
Frequently Bought Together
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Jess or Wes Separation Module, Protein Simple
Supplier: PROTEIN SIMPLE MS
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
AFFINIPURE GOAT ANTI-HUMAN IGG
Supplier: Jackson Immunoresearch Lab