Robotic Pipette Tips
300 µl, +, 96 pieces/box, 10 boxes/pack, 5 packs/case
Catalog # 77507-868
Supplier: NEST SCIENTIFIC
Supplier #: 345609
Jump to:
Product Details & Documents
NEST robotic conductive tip can be applied in R and D of genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals and other high flux liquid transfer.
- Made of polymer material polypropylene (PP)
- Offers high efficiency and repeatability
- With barcode
Surface owns high hydrophobicity as treated with a special technique making for certain purposes.
Specifications
- Volume:300 µl
- Packed:96 pieces/box, 10 boxes/pack, 5 packs/case
- Sterile:+