Disposable Sampler, VTM
Disposable sampler, 3 ml VTM, 10 ml
Catalog # 77507-430
Supplier: NEST SCIENTIFIC
Supplier #: 202117
Jump to:
Product Details & Documents
Disposable sampler viral transport medium.
- Used for collection, storage and transportation of human nasopharyngeal virus samples
- Unused VTM medium can be transported at room temperature
- Sample transport temperature requirement & storage time: 2 to 4 °C, 48 h
Specifications
- Volume:10 ml
- Description:Disposable sampler, 3 ml VTM
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



