Orders LinkContactUsLinkComponent
TNTplus™ Test Kits, COD
20 - 1500 mg/L
  104087-812
Marketsource
 :   Hach
 :  TNT82206
TNTplus™ Test Kits, COD
  104087-812
 :  Hach
Marketsource
 :  
TNTplus™ Test Kits, COD
TNTplus™ Test Kits, COD
20 - 1500 mg/L
 :  
$702.00
Pack of 150
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
  • Range:
    20 - 1500 mg/L