Orders LinkContactUsLinkComponent
TNTplus™ Test Kits, COD
TNTplus™ Test Kits, COD
3 - 150 mg/L
Catalog # 104087-808
Marketsource
Supplier:   Hach
Supplier #:  TNT821
TNTplus™ Test Kits, COD
Catalog # 104087-808
Supplier:  Hach
Marketsource
CAS Number:  
TNTplus™ Test Kits, COD
TNTplus™ Test Kits, COD
3 - 150 mg/L
CAS Number:  
Price
$127.51
Log in for availability
Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
Specifications
Specifications
  • Range:
    3 - 150 mg/L