Orders LinkContactUsLinkComponent
IntelliCAL™ Ion Selective Electrode, Ammonia, Solid-State
IntelliCAL™ Ion Selective Electrode, Ammonia, Solid-State
3 m cable, Non refillable driTEK gel, Porous PTFE annular ring
Catalog # 104087-336
Marketsource
Supplier:   Hach
Supplier #:  ISENH418103
IntelliCAL™ Ion Selective Electrode, Ammonia, Solid-State
Catalog # 104087-336
Supplier:  Hach
Marketsource
IntelliCAL™ Ion Selective Electrode, Ammonia, Solid-State
IntelliCAL™ Ion Selective Electrode, Ammonia, Solid-State
3 m cable, Non refillable driTEK gel, Porous PTFE annular ring
Price
$1,895.54
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
IntelliCAL™ ISENH4181 is a digital combination ammonium ion selective electrode (ISE) with a non refillable double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Provides fast, stable, and accurate response
  • Ultimate traceability in measurement history
  • User alerted when re-calibration is needed
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Ideal for measuring ammonium concentrations in wastewater, drinking water and general water quality applications.
Delivery information: The electrode is supplied with calibration certificate and Basic User Manual.
The analysis package contains the IntelliCAL ISENH418101 electrode, ammonium ionic strength adjustor powder pillows (100), nitrogen-ammonia 10 mg/L standard solution (500 ml), and nitrogen-ammonia 100 mg/L standard solution (500 ml).
Specifications
  • Junction:
    Porous PTFE annular ring
  • Fill solution:
    Non refillable driTEK gel
  • Connector:
    3 m cable
Accessories
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Ammonium ionic strength adjustor (ISA), 100 powder pillows
    Ammonium ionic strength adjustor (ISA), 100 powder pillows
    104086-742
    Determining very low ammonium levels
    Log in for availability
    Each - $133.54