Sample Cuvettes
10 mm, Optical glass
Catalog # 104087-596
Marketsource
Supplier: Hach
Supplier #: LZP045
Jump to:
Product Details & Documents
Sample cuvettes for photometric measurements.
- Available with or without lid
- Available reusable (made from glass) or for single use (made from plastics)
Specifications
- Window material:Optical glass
- Pathlength:10 mm