Order Entry
United States
Orders LinkContactUsLinkComponent
Test Strips

Test Strips

pH, pH 4 - 9, TS
Catalog # 104086-618
Marketsource
Supplier:   Hach
Supplier #:  2745650
Test Strips
Catalog # 104086-618
Supplier:  Hach
Marketsource
CAS Number:  
Test Strips
Test Strips

pH, pH 4 - 9, TS

CAS Number:  
Price
$29.75
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.

  • Portable - ideal for field testing of surface water
  • Easy to use

Documents

Specifications

Specifications

  • Description:
    pH
  • Range:
    pH 4 - 9
  • Type:
    TS

Specifications