Orders LinkContactUsLinkComponent
Test Strips
Copper test strips, 0 - 3 mg/L, TS
Catalog # 104086-610
Marketsource
Supplier:   Hach
 :  2745125
Test Strips
Catalog # 104086-610
Supplier:  Hach
Marketsource
CAS Number:  
Test Strips
Test Strips
Copper test strips, 0 - 3 mg/L, TS
CAS Number:  
Price
$58.63
Log in for availability
Each
Product Details & Documents
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
Specifications
Specifications
  • Description:
    Copper test strips
  • Range:
    0 - 3 mg/L
  • Type:
    TS
Specifications