Orders LinkContactUsLinkComponent
Test Strips
Alkalinity (free and total CaCO₃), 0 - 240 mg/L, TS
  104086-608
Marketsource
 :   Hach
 :  2744850
Test Strips
  104086-608
 :  Hach
Marketsource
 :  
Test Strips
Test Strips
Alkalinity (free and total CaCO₃), 0 - 240 mg/L, TS
 :  
$30.31
Each
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
  • Description:
    Alkalinity (free and total CaCO₃)
  • Range:
    0 - 240 mg/L
  • Type:
    TS