Order Entry
United States
Orders LinkContactUsLinkComponent
 

pH Buffer Solutions, Color Coded

-, 4.01/7.00/10.01
Catalog # 104086-722
Marketsource
Supplier:   Hach
 :  2947600
pH Buffer Solutions, Color Coded
Catalog # 104086-722
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions, Color Coded
pH Buffer Solutions, Color Coded

-, 4.01/7.00/10.01

CAS Number:  
Price
$58.55
Log in for availability
Each
 
Product Details & Documents
  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C

 : Traceable to NIST.

 

Specifications

Specifications

  • pH:
    4.01/7.00/10.01
  • pH accuracy:
    -

Specifications