Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER FLUORIDE ADJUST 454G

BUFFER FLUORIDE ADJUST 454G

Catalog # 104086-438
Marketsource
Supplier:   Hach
Supplier #:  258901
BUFFER FLUORIDE ADJUST 454G
Catalog # 104086-438
Supplier:  Hach
Marketsource
CAS Number:  
BUFFER FLUORIDE ADJUST 454G
CAS Number:  
Price
$220.15
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents