Buffer Powder Pillows
±0.02, Phosphate, 25 °C, Pack of 15, 7, Yellow
104085-938
Marketsource
: Hach
: 2227095
The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.
- Fresh and reliable
- No danger of contamination
- Traceable to NIST standards
: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.
- Color:Yellow
- Packed:Pack of 15
- pH buffer type:Phosphate
- Reference temperature:25 °C
- pH:7
- pH accuracy:±0.02



