Order Entry
United States
Orders LinkContactUsLinkComponent
Buffer Powder Pillows

Buffer Powder Pillows

±0.02, Phosphate, 25 °C, Pack of 250, 7, Yellow
Catalog # 104085-934
Marketsource
Supplier:   Hach
Supplier #:  2227064
Buffer Powder Pillows
Catalog # 104085-934
Supplier:  Hach
Marketsource
CAS Number:  
Buffer Powder Pillows
Buffer Powder Pillows

±0.02, Phosphate, 25 °C, Pack of 250, 7, Yellow

CAS Number:  
Price
$200.77
Log in for availability
Pack of 250
Minimum order quantity is 1
Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

Delivery information: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Specifications

Specifications

  • Color:
    Yellow
  • Packed:
    Pack of 250
  • pH buffer type:
    Phosphate
  • Reference temperature:
    25 °C
  • pH:
    7
  • pH accuracy:
    ±0.02
  • Cat. No.:
    104085-934

Specifications