Orders LinkContactUsLinkComponent
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Chromium, model CH-14, 50 - 1000 mg/L Cr(6+)
Catalog # 104085-932
Marketsource
Supplier:   Hach
Supplier #:  222702
Test Kits, Drop Count Tests
Catalog # 104085-932
Supplier:  Hach
Marketsource
CAS Number:  
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Chromium, model CH-14, 50 - 1000 mg/L Cr(6+)
CAS Number:  
Price
$177.23
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Description:
    Chromium, model CH-14
  • Range:
    50 - 1000 mg/L Cr(6+)
Specifications