Orders LinkContactUsLinkComponent
Test Strips
Test Strips
Chloride, 300 - 6000 mg/L, TS
Catalog # 104086-624
Marketsource
Supplier:   Hach
Supplier #:  2751340
Test Strips
Catalog # 104086-624
Supplier:  Hach
Marketsource
CAS Number:  
Test Strips
Test Strips
Chloride, 300 - 6000 mg/L, TS
CAS Number:  
Price
$124.46
Log in for availability
Pack of 40
Minimum order quantity is 1
Product Details & Documents
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
Documents
Specifications
Specifications
  • Description:
    Chloride
  • Range:
    300 - 6000 mg/L
  • Type:
    TS
Specifications