Orders LinkContactUsLinkComponent
TPH Reagent Sets
Alkaline cyanide, 100 ml, PAN, 0.006 - 0.700 mg/L, -, -, RS
Catalog # 104085-842
Marketsource
Supplier:   Hach
 :  2122332
TPH Reagent Sets
Catalog # 104085-842
Supplier:  Hach
Marketsource
CAS Number:  
TPH Reagent Sets
TPH Reagent Sets
Alkaline cyanide, 100 ml, PAN, 0.006 - 0.700 mg/L, -, -, RS
CAS Number:  
Price
$63.26
Log in for availability
Each
Product Details & Documents
For the determination of total petroleum hydrocarbons (TPH) in soil and water.
 : Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Specifications
Specifications
  • Chemical method:
    PAN
  • Description:
    Alkaline cyanide, 100 ml
  • Range:
    0.006 - 0.700 mg/L
  • Type:
    RS
  • Sample Size:
  • No. of tests:
Specifications