Orders LinkContactUsLinkComponent
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Sodium thiosulfate standard solution, stabilised, 0.00246 N, 500 ml bottle, -
Catalog # 104086-194
Marketsource
Supplier:   Hach
Supplier #:  2408549
Test Kits, Drop Count Tests
Catalog # 104086-194
Supplier:  Hach
Marketsource
CAS Number:  
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Sodium thiosulfate standard solution, stabilised, 0.00246 N, 500 ml bottle, -
CAS Number:  
Price
$67.08
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Description:
    Sodium thiosulfate standard solution, stabilised, 0.00246 N, 500 ml bottle
  • Range:
Specifications